Please login to your online account to display your discounted pricing

5-Bromo-2-fluorotoluene, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  269830050

 View more versions of this product

Catalog No. AC269830050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 189.03
Quantity 5g
Color Clear colorless
EINECS Number 257-202-9
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Name Note 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 5-Bromo-2-fluorotoluene
Assay Percent Range 99%
Molecular Formula C7H6BrF
Linear Formula CH3C6H3BrF
MDL Number MFCD00000343
CAS 51437-00-4
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Bromo-2-fluorotoluene
Name Note 99%
CAS 51437-00-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6BrF
Linear Formula CH3C6H3BrF
Formula Weight 189.03
MDL Number MFCD00000343
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents