Please login to your online account to display your discounted pricing

5-Bromo-2-fluorotoluene, 99%, ACROS Organics™

$56.25 - $56.25

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorotoluene
Name Note 99%
CAS 51437-00-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6BrF
Linear Formula CH3C6H3BrF
Formula Weight 189.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269830250

  SDS
thermo fisher scientific
269830250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC269830050

  SDS
thermo fisher scientific
269830050
Glass bottle 5g Each for $56.25
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 189.03
Quantity 25g
Color Clear colorless
EINECS Number 257-202-9
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-Bromo-2-fluorotoluene
Linear Formula CH3C6H3BrF
CAS Min % 98.5
MDL Number MFCD00000343
CAS 51437-00-4
Assay Percent Range 99%
Molecular Formula C7H6BrF
Name Note 99%
Packaging Glass bottle
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.