Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluorotoluene, 98+%

$53.04 - $620.78

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorotoluene
CAS 51437-00-4
Molecular Formula C7H6BrF
Formula Weight 189.03
MDL Number MFCD00000343
Beilstein 2242693
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1072106

  SDS
alfa aesar™
A1072106
5g Each for $53.04

AAA1072114

  SDS
alfa aesar™
A1072114
25g Each for $179.31

AAA1072122

  SDS
alfa aesar™
A1072122
100g Each for $620.78
Description & Specifications

Specifications

Formula Weight 189.03
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Bromo-2-fluorotoluene
Boiling Point 94° to 95°C (50mmHg)
MDL Number MFCD00000343
Flash Point 73°C (163°F)
Molecular Formula C7H6BrF
Beilstein 2242693
Quantity 5g
CAS 51437-00-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Assay ≥98%

Applications
5-Bromo-2-fluorotoluene is used in the preparation of 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene and 3-(4-fuoro-3-methylphenyl)acrylamide. It is also used as an intermediates of medicine.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.