missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2-fluoropyrimidine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2779503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Bromo-2-fluoropyrimidine | |
90°C to 93°C | |
MFCD07787364 | |
2-fluoro-5-bromopyrimidine, 5-bromo-2-fluoro-pyrimidine, pyrimidine, 5-bromo-2-fluoro, pubchem3589, acmc-1b67r, ksc352k7r, #, 5-bromanyl-2-fluoranyl-pyrimidine, abbypharma ap-11-1804 | |
FC1=NC=C(Br)C=N1 | |
176.98 | |
176.98 |
62802-38-4 | |
C4H2BrFN2 | |
1 g | |
CTWZYPZCDJKBRS-UHFFFAOYSA-N | |
5-bromo-2-fluoropyrimidine | |
600690 | |
95% |
Chemical Identifiers
62802-38-4 | |
176.98 | |
CTWZYPZCDJKBRS-UHFFFAOYSA-N | |
600690 | |
FC1=NC=C(Br)C=N1 |
C4H2BrFN2 | |
MFCD07787364 | |
2-fluoro-5-bromopyrimidine, 5-bromo-2-fluoro-pyrimidine, pyrimidine, 5-bromo-2-fluoro, pubchem3589, acmc-1b67r, ksc352k7r, #, 5-bromanyl-2-fluoranyl-pyrimidine, abbypharma ap-11-1804 | |
5-bromo-2-fluoropyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only