Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluoropyridine-3-boronic acid pinacol ester, 95%

$196.72 - $196.72

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluoropyridine-3-boronic acid pinacol ester
CAS 1073353-50-0
Molecular Formula C11H14BBrFNO2
Formula Weight 301.95g/mol
MDL Number MFCD08689619
Synonym 5-Bromo-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2782003

 
alfa aesar™
H27820-03
1g Each for $196.72
Description & Specifications

Specifications

Melting Point 46° to 49°C
Quantity 1g
Chemical Name or Material 5-Bromo-2-fluoropyridine-3-boronic acid pinacol ester
CAS 1073353-50-0
Assay 95%
Molecular Formula C11H14BBrFNO2
Formula Weight 301.95g/mol
MDL Number MFCD08689619
Synonym 5-Bromo-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.