missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluoropyridine-3-boronic acid, 98%

$114.36 - $1,309.93

Chemical Identifiers

CAS 501435-91-2
Molecular Formula C5H4BBrFNO2
Formula Weight 219.81
MDL Number MFCD04039317
Synonym 5-Bromo-2-fluoro-3-pyridylboronic acid
Chemical Name or Material 5-Bromo-2-fluoropyridine-3-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1991503 SDS Alfa Aesar™
L1991503
1g Each for $338.64
AAL1991506 SDS Alfa Aesar™
L1991506
5g Each for $1,309.93
AAL19915MD SDS Alfa Aesar™
L19915MD
250mg Each for $114.36
Specifications Description & Specifications

Chemical Identifiers

501435-91-2
219.81
5-Bromo-2-fluoro-3-pyridylboronic acid
C5H4BBrFNO2
MFCD04039317
5-Bromo-2-fluoropyridine-3-boronic acid

Specifications

501435-91-2
219.81
>300°C
1g
98%
C5H4BBrFNO2
MFCD04039317
5-Bromo-2-fluoro-3-pyridylboronic acid
5-Bromo-2-fluoropyridine-3-boronic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning