Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluorocinnamic acid, 98%

$66.30 - $244.09

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorocinnamic acid
CAS 202865-71-2
Molecular Formula C9H6BrFO2
Formula Weight 245.05
MDL Number MFCD00143427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1610906

  SDS
alfa aesar™
A1610906
5g Each for $66.30

AAA1610914

  SDS
alfa aesar™
A1610914
25g Each for $244.09
Description & Specifications

Specifications

Formula Weight 245.05
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 193° to 196°C
Chemical Name or Material 5-Bromo-2-fluorocinnamic acid
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C9H6BrFO2
Health Hazard 1 Warning
MDL Number MFCD00143427
CAS 202865-71-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.