Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluorobenzotrifluoride, 98%

$123.91 - $477.18

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorobenzotrifluoride
CAS 393-37-3
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00042493
Beilstein 2643550
Synonym 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene; 4-Fluoro-3-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1673606

 
alfa aesar™
A1673606
5g Each for $123.91

AAA1673614

 
alfa aesar™
A1673614
25g Each for $477.18
Description & Specifications

Specifications

Formula Weight 243
Chemical Name or Material 5-Bromo-2-fluorobenzotrifluoride
Assay 98%
Molecular Formula C7H3BrF4
MDL Number MFCD00042493
CAS 393-37-3
Beilstein 2643550
Synonym 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene; 4-Fluoro-3-(trifluoromethyl)bromobenzene
Boiling Point 79° to 80°C (50mmHg)
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in water 700 g/L (20°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.