5-Bromo-2-fluorobenzotrifluoride, 97%, Acros Organics™
Manufacturer:  Acros Organics AC442890050
 View more versions of this product
Catalog No. AC442890050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
5-Bromo-2-fluorobenzotrifluoride
Authentic
5g
C7H3BrF4
AJLIJYGWAXPEOK-UHFFFAOYSA-N
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
2736325
97%
110°C
142.0°C to 147.0°C
393-37-3
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
C1=CC(=C(C=C1Br)C(F)(F)F)F
242.999
243
Chemical Identifiers
393-37-3
242.999
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
C7H3BrF4
AJLIJYGWAXPEOK-UHFFFAOYSA-N
2736325
C1=CC(=C(C=C1Br)C(F)(F)F)F
Safety and Handling
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Warning
SDS Documents