Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluorobenzamide, 96%

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorobenzamide
CAS 214210-17-0
Formula Weight 218.02
MDL Number MFCD03094430
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6245703

 
alfa aesar™
H6245703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62457MD

 
alfa aesar™
H62457MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.02
CAS 214210-17-0
Chemical Name or Material 5-Bromo-2-fluorobenzamide
Physical Form Solid
Quantity 1g
MDL Number MFCD03094430
Assay 96%