missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-fluorobenzaldehyde, TCI America™

$44.31 - $131.39

Chemical Identifiers

CAS 93777-26-5
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00070755
InChI Key MMFGGDVQLQQQRX-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
PubChem CID 736327
IUPAC Name 5-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17895G View Documents TCI America
B17895G
5g Each for $44.31
 
B178925G View Documents TCI America
B178925G
25g Each for $131.39
 
Specifications

Chemical Identifiers

93777-26-5
203.01
MMFGGDVQLQQQRX-UHFFFAOYSA-N
736327
C1=CC(=C(C=C1Br)C=O)F
C7H4BrFO
MFCD00070755
2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
5-bromo-2-fluorobenzaldehyde

Specifications

White-Yellow
93777-26-5
MFCD00070755
MMFGGDVQLQQQRX-UHFFFAOYSA-N
5-bromo-2-fluorobenzaldehyde
736327
Crystal-Lump at 20°C
5-Bromo-2-fluorobenzaldehyde
5g
C7H4BrFO
2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
C1=CC(=C(C=C1Br)C=O)F
203.01
203.01
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No