Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluorobenzaldehyde, 98%

$232.04 - $961.31

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluorobenzaldehyde
CAS 93777-26-5
Molecular Formula C7H4BrFO
Formula Weight 203.01
MDL Number MFCD00070755
Beilstein 7632798
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1421918

 
alfa aesar™
A1421918
50g Each for $232.04

AAA1421930

 
alfa aesar™
A1421930
250g Each for $961.31

AAA1421909

 
alfa aesar™
A1421909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Bromo-2-fluorobenzaldehyde
CAS 93777-26-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C7H4BrFO
Formula Weight 203.01
MDL Number MFCD00070755
Beilstein 7632798
Boiling Point 230°C
Quantity 50g
Flash Point 109°C (228°F)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.