Learn More
5'-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6128206
Description
5'-Bromo-2'-fluoroacetophenone is an intermediate used in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5′-Bromo-2′-fluoroacetophenone is an intermediate used in pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed.
Specifications
5'-Bromo-2'-fluoroacetophenone | |
C8H6BrFO | |
5 g | |
Slightly soluble in water. | |
CC(=O)C1=C(C=CC(=C1)Br)F | |
217.037 | |
217.04 | |
Solid |
198477-89-3 | |
MFCD11226880 | |
1-5-bromo-2-fluorophenyl ethanone, 1-5-bromo-2-fluorophenyl-1-ethanone, 1-5-bromo-2-fluorophenyl ethan-1-one, ethanone, 1-5-bromo-2-fluorophenyl, 5'-bromo-2'-fluoroacetophenone, 1-acetyl-5-bromo-2-fluorobenzene, 1-5-bromo-2-fluoro-phenyl-ethanone, pubchem16430, acmc-1c1do, 5-bromo-2-fluoroacetophenone | |
XNRQIHIOKXQSPG-UHFFFAOYSA-N | |
1-(5-bromo-2-fluorophenyl)ethanone | |
22607476 | |
98% |
Chemical Identifiers
198477-89-3 | |
217.037 | |
XNRQIHIOKXQSPG-UHFFFAOYSA-N | |
22607476 | |
CC(=O)C1=C(C=CC(=C1)Br)F |
C8H6BrFO | |
MFCD11226880 | |
1-5-bromo-2-fluorophenyl ethanone, 1-5-bromo-2-fluorophenyl-1-ethanone, 1-5-bromo-2-fluorophenyl ethan-1-one, ethanone, 1-5-bromo-2-fluorophenyl, 5'-bromo-2'-fluoroacetophenone, 1-acetyl-5-bromo-2-fluorobenzene, 1-5-bromo-2-fluoro-phenyl-ethanone, pubchem16430, acmc-1c1do, 5-bromo-2-fluoroacetophenone | |
1-(5-bromo-2-fluorophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only