missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluoro-4-methylpyridine, 96%

Manufacturer:  Alfa Aesar™ H6143122

 View more versions of this product

Catalog No. AAH6143122



Description

Description

Applications
5-Bromo-2-fluoro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

5-Bromo-2-fluoro-4-methylpyridine
Colorless to Yellow
864830-16-0
MFCD09839213
PFODFMZCBQUYNG-UHFFFAOYSA-N
5-bromo-2-fluoro-4-methylpyridine
23157062
Liquid
1.5300
100g
C6H5BrFN
2-fluoro-4-methyl-5-bromopyridine
CC1=CC(=NC=C1Br)F
190.015
190.01
96%

Chemical Identifiers

864830-16-0
190.015
PFODFMZCBQUYNG-UHFFFAOYSA-N
23157062
CC1=CC(=NC=C1Br)F
C6H5BrFN
MFCD09839213
2-fluoro-4-methyl-5-bromopyridine
5-bromo-2-fluoro-4-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents