missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-fluoro-3-methylpyridine, TCI America™

$61.11 - $181.81

Chemical Identifiers

CAS 29312-98-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD03095091
InChI Key JLCVSFDCHKCISN-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-3-picoline, 2-fluoro-5-bromo-3-methylpyridine, pyridine, 5-bromo-2-fluoro-3-methyl, 2-fluoro-5-bromo-3-picoline, 2-fluoro-3-methyl-5-bromopyridine, 5-bromo-2-fluoro-3-methyl pyridine, pubchem1136, acmc-1cq2z, ksc495o9n, 5-bromo-2-fluoro-3-methyl-pyridine
PubChem CID 12056902
IUPAC Name 5-bromo-2-fluoro-3-methylpyridine
SMILES CC1=CC(=CN=C1F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37751G View Documents TCI America
B37751G
1g Each for $61.11
 
B37755G View Documents TCI America
B37755G
5g Each for $181.81
 
Specifications

Chemical Identifiers

29312-98-9
190.015
JLCVSFDCHKCISN-UHFFFAOYSA-N
12056902
CC1=CC(=CN=C1F)Br
C6H5BrFN
MFCD03095091
5-bromo-2-fluoro-3-picoline, 2-fluoro-5-bromo-3-methylpyridine, pyridine, 5-bromo-2-fluoro-3-methyl, 2-fluoro-5-bromo-3-picoline, 2-fluoro-3-methyl-5-bromopyridine, 5-bromo-2-fluoro-3-methyl pyridine, pubchem1136, acmc-1cq2z, ksc495o9n, 5-bromo-2-fluoro-3-methyl-pyridine
5-bromo-2-fluoro-3-methylpyridine

Specifications

Undesignated
1g
C6H5BrFN
5-bromo-2-fluoro-3-picoline, 2-fluoro-5-bromo-3-methylpyridine, pyridine, 5-bromo-2-fluoro-3-methyl, 2-fluoro-5-bromo-3-picoline, 2-fluoro-3-methyl-5-bromopyridine, 5-bromo-2-fluoro-3-methyl pyridine, pubchem1136, acmc-1cq2z, ksc495o9n, 5-bromo-2-fluoro-3-methyl-pyridine
CC1=CC(=CN=C1F)Br
190.015
190.02
≥98.0% (GC)
65°C
29312-98-9
MFCD03095091
JLCVSFDCHKCISN-UHFFFAOYSA-N
5-bromo-2-fluoro-3-methylpyridine
12056902
Crystal-Powder at 20°C
5-Bromo-2-fluoro-3-methylpyridine
Safety and Handling

Safety and Handling

TSCA : No