Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-fluoro-3-methylpyridine, 95%

$140.74 - $140.74

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-fluoro-3-methylpyridine
CAS 29312-98-9
Molecular Formula C6H5BrFN
Formula Weight 190.01
MDL Number MFCD03095091
Synonym 5-Bromo-2-fluoro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2613103

  SDS
alfa aesar™
H2613103
1g Each for $140.74
Description & Specifications

Specifications

Formula Weight 190.01
Chemical Name or Material 5-Bromo-2-fluoro-3-methylpyridine
Assay 95%
Molecular Formula C6H5BrFN
MDL Number MFCD03095091
CAS 29312-98-9
Melting Point 60° to 63°C
Synonym 5-Bromo-2-fluoro-3-picoline
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.