Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-ethoxypyridine, 97%

$50.62 - $485.18

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-ethoxypyridine
CAS 55849-30-4
Molecular Formula C7H8BrNO
Formula Weight 202.05
MDL Number MFCD00234311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2719703

  SDS
alfa aesar™
H2719703
1g Each for $50.62

AAH2719706

  SDS
alfa aesar™
H2719706
5g Each for $156.70

AAH2719714

  SDS
alfa aesar™
H2719714
25g Each for $485.18
Description & Specifications

Specifications

Formula Weight 202.05
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Chemical Name or Material 5-Bromo-2-ethoxypyridine
Melting Point 32° to 36°C
Molecular Formula C7H8BrNO
Quantity 1g
MDL Number MFCD00234311
Health Hazard 1 Danger
CAS 55849-30-4
Assay 97%
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.