Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-ethoxybenzaldehyde, 98%

$223.00 - $223.00

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-ethoxybenzaldehyde
CAS 79636-94-5
Molecular Formula C9H9BrO2
Formula Weight 229.08
MDL Number MFCD00016600
Beilstein 3244183
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1762114

  SDS
alfa aesar™
A1762114
25g Each for $223.00
Description & Specifications

Specifications

Formula Weight 229.08
Melting Point 67° to 71°C
Beilstein 3244183
Molecular Formula C9H9BrO2
CAS 79636-94-5
MDL Number MFCD00016600
Assay 98%
Quantity 25g
Chemical Name or Material 5-Bromo-2-ethoxybenzaldehyde