Please login to your online account to display your discounted pricing

5-Bromo-2«-deoxyuridine, Fisher BioReagents


Quantity 250mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2508250

 View more versions of this product

Catalog No. BP2508250

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 307.1
Melting Point 191°C
Quantity 250mg
Merck Index 15, 1461
Molecular Formula C9H11BrN2O5
Color Off-white
Chemical Name or Material 5-Bromo-2'-Deoxyuridine
Health Hazard 1 Signal Word:WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes and skin. Possible risks of irreversible effects.
NFPA
Health:1 Flammability:0 Instability:0
Identification Pass Test
Recommended Storage -20°C
Packaging Amber Glass
Name Note Off-white Powder
Assay Percent Range >99%
CAS 59-14-3
ChemAlert Storage Symbol Gray
Synonym 5- Bromouracil-2-Deoxyriboside
Lambda Max 276 to 282nm
Infrared Spectrum Conforms to reference

Specifications

Chemical Name or Material 5-Bromo-2'-Deoxyuridine
Name Note Off-white Powder
CAS 59-14-3
Assay Percent Range >99%
Molecular Formula C9H11BrN2O5
Formula Weight 307.1
Synonym 5- Bromouracil-2-Deoxyriboside
Accessories
 

Signal Word:WARNING!

Emergency Overview
Irritating to eyes and skin. Possible risks of irreversible effects.
NFPA
Health:1 Flammability:0 Instability:0

-20°C

SDS & Safety Documents