missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2«-deoxyuridine, Fisher BioReagents
Manufacturer: Fisher BioReagents BP25085
Description
5-Bromo-2'-Deoxyuridine is a thymidine analog used as a mutagen in genetic research.Specifications
5-Bromo-2'-Deoxyuridine | |
59-14-3 | |
191°C | |
>99% | |
C9H11BrN2O5 | |
15, 1461 | |
5g | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O | |
307.1 | |
CHEBI:472552 | |
Powder/Solid |
Off-white Powder | |
White | |
Pass Test | |
Conforms to reference | |
Amber Glass | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
6035 | |
307.1 |
Chemical Identifiers
59-14-3 | |
307.1 | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
CHEBI:472552 | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O |
C9H11BrN2O5 | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
6035 | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
Safety and Handling

Emergency Overview
Irritating to eyes and skin. Possible risks of irreversible effects.
NFPA
Health:1 Flammability:0 Instability:0
Signal Word: WARNING!
Recommended Storage : -20°C