missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2'-Deoxyuridine, >99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210017101
Specifications
5-Bromo-2'-Deoxyuridine | |
1g | |
>99% | |
MFCD00006529 | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
6035 | |
307.1 |
White | |
59-14-3 | |
C9H11BrN2O5 | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O | |
307.1 | |
CHEBI:472552 | |
Solid |
Chemical Identifiers
59-14-3 | |
307.1 | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
6035 | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
C9H11BrN2O5 | |
MFCD00006529 | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
CHEBI:472552 | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O |
Safety and Handling
EINECSNumber : 200-415-9
Recommended Storage : Store at -20°C.