Learn More
5-Bromo-2'-deoxyuridine, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 228590025
Specifications
0.47 to 0.53 (A250/A260),1.87 to 1.93 (A280/A260),1.45 to 1.55 (A290/A260) | |
Authentic | |
1% max. (HPLC) Total impurities | |
+ 31.00 | |
White to Yellow | |
2.5g | |
99.0 | |
99% min. (HPLC) | |
MFCD00006529 | |
15, 1461 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,dmso (50mg/mL) | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O | |
307.09 | |
CHEBI:472552 | |
Crystals or Crystalline Powder |
5-Bromo-2′-deoxyuridine | |
Glass bottle | |
0.2% max. ethanol (GC), 0.1% max. methanol (GC) | |
2% max. (K.F.) | |
191.0°C to 194.0°C | |
59-14-3 | |
100.0 | |
C9H11BrN2O5 | |
24, IV, 1234 | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
6035 | |
307.09 | |
99+% |
Chemical Identifiers
59-14-3 | |
307.09 | |
WOVKYSAHUYNSMH-RRKCRQDMSA-N | |
6035 | |
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
C9H11BrN2O5 | |
MFCD00006529 | |
5-brdu, 5-bromo-2'-deoxyuridine, 5-bromodeoxyuridine, 5-bromodesoxyuridine, 5-bromouracil deoxyriboside, brdu, bromodeoxyuridine, bromouracil deoxyriboside, broxuridine, budr | |
CHEBI:472552 | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Br)CO)O |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-415-9