Please login to your online account to display your discounted pricing

5-Bromo-2'-deoxycytidine, MP Biomedicals™


5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP

Quantity 1g

Manufacturer: mp biomedicals  0210016691

 View more versions of this product

Catalog No. ICN10016691

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material 5-Bromo-2'-deoxycytidine monohydrate
CAS 1022-79-3
Physical Form White powder
Molecular Formula C9H12BrN3O4 · ×H2O
Formula Weight 306.1g/mol (anhydrous)
Synonym 5-Bromo-2'-Deoxycytidine
Recommended Storage Store at 0°C. Store Desiccated.
EINECS Number 213-824-2

Specifications

Chemical Name or Material 5-Bromo-2'-deoxycytidine monohydrate
CAS 1022-79-3
Physical Form White powder
Molecular Formula C9H12BrN3O4 · ×H2O
Formula Weight 306.1g/mol (anhydrous)
Synonym 5-Bromo-2'-Deoxycytidine
Accessories

Store at 0°C. Store Desiccated.