missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-cyanopyridine, TCI America™

$59.58 - $180.28

Chemical Identifiers

CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B36105G View Documents TCI America
B36105G
5g Each for $59.58
 
B361025G View Documents TCI America
B361025G
25g Each for $180.28
 
Specifications

Chemical Identifiers

97483-77-7
183.008
DMSHUVBQFSNBBL-UHFFFAOYSA-N
817154
C1=CC(=NC=C1Br)C#N
C6H3BrN2
MFCD00234144
5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
5-bromopyridine-2-carbonitrile

Specifications

110°C
131°C
97483-77-7
MFCD00234144
5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
C1=CC(=NC=C1Br)C#N
183.008
183.01
≥98.0% (GC)
Undesignated
5g
C6H3BrN2
3439
DMSHUVBQFSNBBL-UHFFFAOYSA-N
5-bromopyridine-2-carbonitrile
817154
Crystal-Powder at 20°C
5-Bromo-2-cyanopyridine
Safety and Handling

Safety and Handling

TSCA : No