Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-cyanobenzeneboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-cyanobenzeneboronic acid
CAS 1032231-30-3
Formula Weight 225.84g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6127303

 
alfa aesar™
H61273-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61273MD

 
alfa aesar™
H61273-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 5-Bromo-2-cyanobenzeneboronic acid
CAS 1032231-30-3
Physical Form Solid
Assay 97%
Formula Weight 225.84g/mol