missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1060214

 View more versions of this product

Catalog No. AAA1060214



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromo-2-chlorotoluene
1.549
25 g
C7H6BrCl
3234896
OZFQMHJKAODEON-UHFFFAOYSA-N
4-bromo-1-chloro-2-methylbenzene
609899
98%
54932-72-8
90°C to 93°C (10 mmHg)
1.5745
MFCD00018503
5-bromo-2-chlorotoluene, benzene, 4-bromo-1-chloro-2-methyl, 2-chloro-5-bromotoluene, pubchem3604, 5-bromo-2-chlorotoulene, acmc-209lk8, ksc493o2d, ozfqmhjkaodeon-uhfffaoysa, attercop-chm at115283
CC1=C(C=CC(=C1)Br)Cl
205.479
205.48

Chemical Identifiers

54932-72-8
205.479
OZFQMHJKAODEON-UHFFFAOYSA-N
609899
CC1=C(C=CC(=C1)Br)Cl
C7H6BrCl
MFCD00018503
5-bromo-2-chlorotoluene, benzene, 4-bromo-1-chloro-2-methyl, 2-chloro-5-bromotoluene, pubchem3604, 5-bromo-2-chlorotoulene, acmc-209lk8, ksc493o2d, ozfqmhjkaodeon-uhfffaoysa, attercop-chm at115283
4-bromo-1-chloro-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only