Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloropyrimidine, 98+%


Quantity 1g

Manufacturer: alfa aesar™  L1836003

 View more versions of this product

Catalog No. AAL1836003

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 193.43
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00483232
Assay ≥98%
Molecular Formula C4H2BrClN2
CAS 32779-36-5
Melting Point 77° to 80°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 507955
Quantity 1g
Boiling Point 95°C (15mmHg)
Chemical Name or Material 5-Bromo-2-chloropyrimidine

Specifications

Chemical Name or Material 5-Bromo-2-chloropyrimidine
CAS 32779-36-5
Molecular Formula C4H2BrClN2
Formula Weight 193.43
MDL Number MFCD00483232
Beilstein 507955

Applications
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.

Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents