Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloropyridine-3-boronic acid, 95%

$172.41 - $172.41

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-chloropyridine-3-boronic acid
CAS 1072944-19-4
Molecular Formula C5H4BBrClNO2
Formula Weight 236.26
MDL Number MFCD09037486
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26966MD

  SDS
alfa aesar™
H26966MD
250mg Each for $172.41
Description & Specifications

Specifications

Formula Weight 236.26
Chemical Name or Material 5-Bromo-2-chloropyridine-3-boronic acid
Assay 95%
Molecular Formula C5H4BBrClNO2
MDL Number MFCD09037486
CAS 1072944-19-4
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.