Please login to your online account to display your discounted pricing

5-Bromo-2-chlorobenzotrifluoride, 99%, ACROS Organics™


Quantity 5g

Manufacturer: fisher scientific acros  165910050

 View more versions of this product

Catalog No. AC165910050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥98.5%
Formula Weight 259.44
Quantity 5g
Color Clear pale yellow
EINECS Number 207-149-2
CAS Max % 100.0
Chemical Name or Material 5-Bromo-2-chlorobenzotrifluoride
Name Note tech., 94%
Assay Percent Range 99%
Molecular Formula C7H3BrClF3
Linear Formula BrC6H3(Cl)CF3
MDL Number MFCD00000601
CAS 445-01-2
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 5-Bromo-2-chloro-α, α, α-trifluorotoluene
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Glass bottle
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)

Specifications

Chemical Name or Material 5-Bromo-2-chlorobenzotrifluoride
Name Note tech., 94%
CAS 445-01-2
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H3BrClF3
Linear Formula BrC6H3(Cl)CF3
Formula Weight 259.44
MDL Number MFCD00000601
Synonym 5-Bromo-2-chloro-α, α, α-trifluorotoluene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents