missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2-chlorobenzonitrile 98.0+%, TCI America™
$190.78 - $715.03
Chemical Identifiers
CAS | 57381-44-9 |
---|---|
Molecular Formula | C7H3BrClN |
Molecular Weight (g/mol) | 216.462 |
MDL Number | MFCD00672948 |
InChI Key | NKDMQBUJOXQVEA-UHFFFAOYSA-N |
PubChem CID | 21525431 |
IUPAC Name | 5-bromo-2-chlorobenzonitrile |
SMILES | C1=CC(=C(C=C1Br)C#N)Cl |
Chemical Identifiers
57381-44-9 | |
216.462 | |
NKDMQBUJOXQVEA-UHFFFAOYSA-N | |
5-bromo-2-chlorobenzonitrile |
C7H3BrClN | |
MFCD00672948 | |
21525431 | |
C1=CC(=C(C=C1Br)C#N)Cl |
Specifications
57381-44-9 | |
White-Yellow | |
MFCD00672948 | |
3439 | |
C1=CC(=C(C=C1Br)C#N)Cl | |
216.462 | |
216.46 | |
Crystalline Powder |
133°C | |
C7H3BrClN | |
5 g | |
NKDMQBUJOXQVEA-UHFFFAOYSA-N | |
5-bromo-2-chlorobenzonitrile | |
21525431 | |
≥98.0% (GC) | |
5-Bromo-2-chlorobenzonitrile |
Safety and Handling
TSCA : No