Learn More
5-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals
$78.08 - $980.05
Chemical Identifiers
CAS | 21739-92-4 |
---|---|
Molecular Formula | C7H4BrClO2 |
Molecular Weight (g/mol) | 235.461 |
MDL Number | MFCD00002415 |
InChI Key | FGERXQWKKIVFQG-UHFFFAOYSA-N |
Synonym | 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r |
PubChem CID | 33127 |
IUPAC Name | 5-bromo-2-chlorobenzoic acid |
SMILES | C1=CC(=C(C=C1Br)C(=O)O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1221809
|
Thermo Scientific Chemicals
A1221809 |
10 g |
Each for $78.08
|
|
|||||
AAA1221818
|
Thermo Scientific Chemicals
A1221818 |
50 g |
Each for $231.71
|
|
|||||
AAA1221830
|
Thermo Scientific Chemicals
A1221830 |
250 g |
Each for $980.05
|
|
|||||
Description
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.
Solubility
Soluble in water 2.63 g/L @20°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
21739-92-4 | |
235.461 | |
FGERXQWKKIVFQG-UHFFFAOYSA-N | |
33127 | |
C1=CC(=C(C=C1Br)C(=O)O)Cl |
C7H4BrClO2 | |
MFCD00002415 | |
2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r | |
5-bromo-2-chlorobenzoic acid |
Specifications
21739-92-4 | |
Odorless | |
MFCD00002415 | |
2691432 | |
Soluble in water 2.63g/L @20°C. | |
C1=CC(=C(C=C1Br)C(=O)O)Cl | |
235.461 | |
235.47 | |
5-Bromo-2-chlorobenzoic acid |
157°C to 162°C | |
C7H4BrClO2 | |
10 g | |
2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r | |
FGERXQWKKIVFQG-UHFFFAOYSA-N | |
5-bromo-2-chlorobenzoic acid | |
33127 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 244-558-5
TSCA : Yes
RUO – Research Use Only