Please login to your online account to display your discounted pricing

5-Bromo-2-chloroanisole, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398900010

 View more versions of this product

Catalog No. AC398900010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid or Low Melting Solid
Assay ≥97.5%
Formula Weight 221.48
Quantity 1g
Color Clear colorless to pale yellow
Density 1.6310g/mL
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H3(Cl)OCH3
CAS 16817-43-9
CAS Min % 97.5
Health Hazard 1 Warning
Molecular Formula C7H6BrClO
Chemical Name or Material 5-Bromo-2-chloroanisole
MDL Number MFCD00078659
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromo-2-chloroanisole
CAS 16817-43-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrClO
Linear Formula BrC6H3(Cl)OCH3
Formula Weight 221.48
MDL Number MFCD00078659
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents