missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloroanisole, 97%

Manufacturer:  Alfa Aesar™ H55533MD

 View more versions of this product

Catalog No. AAH55533MD



Description

Description

Applications
5-Bromo-2-chloroanisole is used to produce 4-chloro-3-methoxy-benzonitrile at the temperature of 110°C. It will need solvent dimethylformamide

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Bromo-2-chloroanisole
113°C (235°F)
240°C to 245°C
250mg
C7H6BrClO
2-chloro-5-bromoanisole
UAMVKOTWSHJOSY-UHFFFAOYSA-N
4-bromo-1-chloro-2-methoxybenzene
2769567
97%
1.631
1.587
27°C to 28°C
16817-43-9
MFCD00078659
Soluble in water.
COC1=C(C=CC(=C1)Br)Cl
221.478
221.48

Chemical Identifiers

16817-43-9
221.478
UAMVKOTWSHJOSY-UHFFFAOYSA-N
2769567
COC1=C(C=CC(=C1)Br)Cl
C7H6BrClO
MFCD00078659
2-chloro-5-bromoanisole
4-bromo-1-chloro-2-methoxybenzene
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents