missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-chloroanisole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5553303

 View more versions of this product

Catalog No. AAH5553303



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-chloroanisole is used to produce 4-chloro-3-methoxy-benzonitrile at the temperature of 110°C. It will need solvent dimethylformamide

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Bromo-2-chloroanisole
27°C to 28°C
240°C to 245°C
C7H6BrClO
MFCD00078659
5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
UAMVKOTWSHJOSY-UHFFFAOYSA-N
4-bromo-1-chloro-2-methoxybenzene
2769567
97%
16817-43-9
1.631
113°C (235°F)
1.587
1 g
Soluble in water.
COC1=C(Cl)C=CC(Br)=C1
221.48
221.48

Chemical Identifiers

16817-43-9
221.48
UAMVKOTWSHJOSY-UHFFFAOYSA-N
2769567
COC1=C(Cl)C=CC(Br)=C1
C7H6BrClO
MFCD00078659
5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq
4-bromo-1-chloro-2-methoxybenzene
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions