missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloroaniline, 98%

Manufacturer:  Alfa Aesar™ H6410422

 View more versions of this product

Catalog No. AAH6410422



Description

Description

Applications
5-Bromo-2-chloroaniline is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

5-Bromo-2-chloroaniline
100g
C6H5BrClN
UN2811
2-chloro-5-bromoaniline, 3-bromo-6-chloroaniline, 5-bromo-2-chlorophenylamine, benzenamine, 5-bromo-2-chloro, 5-bromo-2-chloro-aniline, 5-bromo-2-chloro-phenylamine, 5-bromo-2-chloro aniline, pubchem3132, 5-bromo-2-chlorobenzenamine, ksc493o4j
C1=CC(=C(C=C1Br)N)Cl
206.467
206.47
98%
145°C to 150°C
60811-17-8
MFCD06796283
2716274
UGOLEPGQWYPIBR-UHFFFAOYSA-N
5-bromo-2-chloroaniline
3830731
Solid

Chemical Identifiers

60811-17-8
206.467
UGOLEPGQWYPIBR-UHFFFAOYSA-N
3830731
C1=CC(=C(C=C1Br)N)Cl
C6H5BrClN
MFCD06796283
2-chloro-5-bromoaniline, 3-bromo-6-chloroaniline, 5-bromo-2-chlorophenylamine, benzenamine, 5-bromo-2-chloro, 5-bromo-2-chloro-aniline, 5-bromo-2-chloro-phenylamine, 5-bromo-2-chloro aniline, pubchem3132, 5-bromo-2-chlorobenzenamine, ksc493o4j
5-bromo-2-chloroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents