missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloro-3-nitropyridine, 98%

$82.05 - $299.62

Chemical Identifiers

CAS 67443-38-3
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.437
MDL Number MFCD00222270
InChI Key WWQQPSDIIVXFOX-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-bromopyridine, 2-chloro-5-bromo-3-nitropyridine, 5-bromo-2-chloro-3-nitro-pyridine, 5-bromo-2-chloro-3-nitropyridin, abbypharma ap-12-5159, acmc-209nz1, ksc352q2l, pubchem1166, pyridine, 5-bromo-2-chloro-3-nitro, zlchem 393
PubChem CID 7019412
IUPAC Name 5-bromo-2-chloro-3-nitropyridine
SMILES C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6489714 View Documents Alfa Aesar™
H6489714
25g Each for $82.05
 
AAH6489722 View Documents Alfa Aesar™
H6489722
100g Each for $299.62
 
Description

Description

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

67443-38-3
237.437
WWQQPSDIIVXFOX-UHFFFAOYSA-N
7019412
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
C5H2BrClN2O2
MFCD00222270
2-chloro-3-nitro-5-bromopyridine, 2-chloro-5-bromo-3-nitropyridine, 5-bromo-2-chloro-3-nitro-pyridine, 5-bromo-2-chloro-3-nitropyridin, abbypharma ap-12-5159, acmc-209nz1, ksc352q2l, pubchem1166, pyridine, 5-bromo-2-chloro-3-nitro, zlchem 393
5-bromo-2-chloro-3-nitropyridine

Specifications

Yellow
25g
C5H2BrClN2O2
UN2811
2-chloro-3-nitro-5-bromopyridine, 2-chloro-5-bromo-3-nitropyridine, 5-bromo-2-chloro-3-nitro-pyridine, 5-bromo-2-chloro-3-nitropyridin, abbypharma ap-12-5159, acmc-209nz1, ksc352q2l, pubchem1166, pyridine, 5-bromo-2-chloro-3-nitro, zlchem 393
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
237.437
237.44
98%
66°C to 68°C
67443-38-3
MFCD00222270
147551
WWQQPSDIIVXFOX-UHFFFAOYSA-N
5-bromo-2-chloro-3-nitropyridine
7019412
Crystalline
5-Bromo-2-chloro-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures