missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloro-3-nitropyridine, 98%

Manufacturer:  Alfa Aesar™ H6489714

 View more versions of this product

Catalog No. AAH6489714



Description

Description

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Specifications

5-Bromo-2-chloro-3-nitropyridine
66°C to 68°C
67443-38-3
MFCD00222270
147551
WWQQPSDIIVXFOX-UHFFFAOYSA-N
5-bromo-2-chloro-3-nitropyridine
7019412
Crystalline
Yellow
25g
C5H2BrClN2O2
UN2811
2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
237.437
237.44
98%

Chemical Identifiers

67443-38-3
237.437
WWQQPSDIIVXFOX-UHFFFAOYSA-N
7019412
C1=C(C(=NC=C1Br)Cl)[N+](=O)[O-]
C5H2BrClN2O2
MFCD00222270
2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
5-bromo-2-chloro-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents