Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloro-3-methylpyridine, 98%

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-chloro-3-methylpyridine
CAS 29241-60-9
Formula Weight 206.47
MDL Number MFCD03095093
Beilstein 1562796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6410906

 
alfa aesar™
H6410906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6410914

 
alfa aesar™
H6410914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.47
Melting Point 40° to 44°C
Chemical Name or Material 5-Bromo-2-chloro-3-methylpyridine
Physical Form Solid
Assay 98%
MDL Number MFCD03095093
CAS 29241-60-9
Beilstein 1562796
Quantity 5g