missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-chloro-3-fluoropyridine, 98%

Manufacturer:  Alfa Aesar™ H6111606

 View more versions of this product

Catalog No. AAH6111606



Specifications

Specifications

5-Bromo-2-chloro-3-fluoropyridine
50°C to 52°C
831203-13-5
MFCD03411574
OEKXUEWGXWCENG-UHFFFAOYSA-N
5-bromo-2-chloro-3-fluoropyridine
2763285
Solid
White
5g
C5H2BrClFN
2-chloro-3-fluoro-5-bromopyridine
C1=C(C(=NC=C1Br)Cl)F
210.43
210.43
98%

Chemical Identifiers

831203-13-5
210.43
OEKXUEWGXWCENG-UHFFFAOYSA-N
2763285
C1=C(C(=NC=C1Br)Cl)F
C5H2BrClFN
MFCD03411574
2-chloro-3-fluoro-5-bromopyridine
5-bromo-2-chloro-3-fluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

Documents