Please login to your online account to display your discounted pricing

5-Bromo-2,4-dimethyl-1,3-thiazole, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC05810DA

 View more versions of this product

Catalog No. CC05810DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.08
Quantity 1g
Assay 97%
Chemical Name or Material 5-Bromo-2, 4-dimethyl-1, 3-thiazole
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5 H6 Br N S
CAS 28599-52-2

Specifications

Chemical Name or Material 5-Bromo-2, 4-dimethyl-1, 3-thiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C5 H6 Br N S
Formula Weight 192.08