Please login to your online account to display your discounted pricing

5-Bromo-2,4-dimethyl-1,3-thiazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromo-2, 4-dimethyl-1, 3-thiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C5 H6 Br N S
Formula Weight 192.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC05810DA

  SDS
fisher scientific company
CC05810DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.08
Quantity 1g
Assay 97%
Chemical Name or Material 5-Bromo-2, 4-dimethyl-1, 3-thiazole
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5 H6 Br N S
CAS 28599-52-2