Please login to your online account to display your discounted pricing

5-Bromo-2,4-dihydroxybenzoic acid monohydrate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Bromo-2, 4-dihydroxybenzoic acid monohydrate
CAS 160348-98-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula C7H5BrO4·H2O
Linear Formula BrC6H2(OH)2CHO
Formula Weight 251.03
MDL Number MFCD00149094
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381930050

  SDS
thermo fisher scientific
AC381930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (on dry substance)
Physical Form Powder
Formula Weight 251.03
Quantity 5g
Color Off-white to pale yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5BrO4·H2O
Linear Formula BrC6H2(OH)2CHO
Packaging Glass bottle
Water 7.8% max.
MDL Number MFCD00149094
CAS 160348-98-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Chemical Name or Material 5-Bromo-2, 4-dihydroxybenzoic acid monohydrate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.