Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2,4-dichloropyrimidine, 98%

Chemical Identifiers

Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine
CAS 36082-50-5
Molecular Formula C4HBrCl2N2
Formula Weight 227.88
MDL Number MFCD00127818
UN Number UN2923
Beilstein 124441
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2536503

 
alfa aesar™
B2536503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2536506

 
alfa aesar™
B2536506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.88
Chemical Name or Material 5-Bromo-2,4-dichloropyrimidine
Assay 98%
Molecular Formula C4HBrCl2N2
MDL Number MFCD00127818
CAS 36082-50-5
UN Number UN2923
Melting Point 29° to 30°C
Beilstein 124441
Boiling Point 128°C (15mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.