missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2,4-di-(tert-butoxy)pyrimidine, 95%

$78.39 - $176.51

Chemical Identifiers

CAS 19752-61-5
Molecular Formula C12H19BrN2O2
Formula Weight 303.2
MDL Number MFCD01927527
Chemical Name or Material 5-Bromo-2,4-di-(tert-butoxy)pyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2754603 SDS Alfa Aesar™
H2754603
1g Each for $176.51
AAH27546MD SDS Alfa Aesar™
H27546MD
250mg Each for $78.39
Specifications Description & Specifications

Chemical Identifiers

19752-61-5
303.2
5-Bromo-2,4-di-(tert-butoxy)pyrimidine
C12H19BrN2O2
MFCD01927527

Specifications

19752-61-5
303.2
58° to 61°C
5-Bromo-2,4-di-(tert-butoxy)pyrimidine
C12H19BrN2O2
MFCD01927527
1g
95%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning