missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic Acid 97.0+%, TCI America™
$245.54 - $927.38
Chemical Identifiers
CAS | 41177-72-4 |
---|---|
Molecular Formula | C9H7BrO3 |
Molecular Weight (g/mol) | 243.06 |
MDL Number | MFCD00191391 |
InChI Key | LEBMKAXASFPSFA-UHFFFAOYSA-N |
Synonym | 5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid |
PubChem CID | 2794797 |
IUPAC Name | 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid |
SMILES | OC(=O)C1=CC(Br)=CC2=C1OCC2 |
Chemical Identifiers
41177-72-4 | |
243.06 | |
LEBMKAXASFPSFA-UHFFFAOYSA-N | |
2794797 | |
OC(=O)C1=CC(Br)=CC2=C1OCC2 |
C9H7BrO3 | |
MFCD00191391 | |
5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid |
Specifications
41177-72-4 | |
C9H7BrO3 | |
1 g | |
LEBMKAXASFPSFA-UHFFFAOYSA-N | |
5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
2794797 | |
≥97.0% (T) | |
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic Acid |
232°C | |
MFCD00191391 | |
5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
OC(=O)C1=CC(Br)=CC2=C1OCC2 | |
243.06 | |
243.06 | |
Crystalline Powder |
Safety and Handling
TSCA : No