Please login to your online account to display your discounted pricing

5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO07141DA

 View more versions of this product

Catalog No. MO07141DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 297.56
Quantity 1g
Assay 97%
CAS 690632-00-9
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-sulfonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H6BrClO3S

Specifications

Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6BrClO3S
Formula Weight 297.56