Please login to your online account to display your discounted pricing

5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6BrClO3S
Formula Weight 297.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07141DA

  SDS
fisher scientific company
MO07141DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO07141DE

  SDS
fisher scientific company
MO07141DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.56
Quantity 1g
Assay 97%
CAS 690632-00-9
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-sulfonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H6BrClO3S