missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2,3-difluoropyridine, 97%

$44.09 - $723.26

Chemical Identifiers

CAS 89402-44-8
Molecular Formula C5H2BrF2N
Molecular Weight (g/mol) 193.979
MDL Number MFCD06659525
InChI Key QVIQXJRQVOPYGI-UHFFFAOYSA-N
Synonym pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine
PubChem CID 14549654
IUPAC Name 5-bromo-2,3-difluoropyridine
SMILES C1=C(C(=NC=C1Br)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6456603 View Documents Alfa Aesar™
H6456603
1g Each for $44.09
 
AAH6456606 View Documents Alfa Aesar™
H6456606
5g Each for $174.26
 
AAH6456614 View Documents Alfa Aesar™
H6456614
25g Each for $723.26
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

89402-44-8
193.979
QVIQXJRQVOPYGI-UHFFFAOYSA-N
14549654
C1=C(C(=NC=C1Br)F)F
C5H2BrF2N
MFCD06659525
pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine
5-bromo-2,3-difluoropyridine

Specifications

Colorless to Yellow
89402-44-8
MFCD06659525
pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine
C1=C(C(=NC=C1Br)F)F
193.979
193.98
97%
1g
C5H2BrF2N
13412561
QVIQXJRQVOPYGI-UHFFFAOYSA-N
5-bromo-2,3-difluoropyridine
14549654
Liquid
5-Bromo-2,3-difluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning