Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2,3-difluorophenol, 97%

$38.12 - $38.12

Chemical Identifiers

Chemical Name or Material 5-Bromo-2,3-difluorophenol
CAS 186590-26-1
Molecular Formula C6H3BrF2O
Formula Weight 208.99
MDL Number MFCD01631350
Beilstein 8831044
Synonym 3-Bromo-5,6-difluorophenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2385303

  SDS
alfa aesar™
B2385303
1g Each for $38.12
Description & Specifications

Specifications

Formula Weight 208.99
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Bromo-2,3-difluorophenol
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 186590-26-1
Beilstein 8831044
MDL Number MFCD01631350
Molecular Formula C6H3BrF2O
Quantity 1g
Synonym 3-Bromo-5,6-difluorophenol
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.