Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2,3-difluoroanisole, 97%

$286.28 - $286.28

Chemical Identifiers

Chemical Name or Material 5-Bromo-2,3-difluoroanisole
CAS 261762-35-0
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD01631352
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2365509

  SDS
alfa aesar™
B2365509
10g Each for $286.28
Description & Specifications

Specifications

Formula Weight 223.02
Melting Point 33° to 36°C
Quantity 10g
MDL Number MFCD01631352
Chemical Name or Material 5-Bromo-2,3-difluoroanisole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Molecular Formula C7H5BrF2O
CAS 261762-35-0
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.